Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-6-(delta-2-Isopentenyl)adenosine hemihydrate, 99+%, Thermo Scientific™

Catalog No. AC226481000
Click to view available options
Quantity:
100 mg

Chemical Identifiers

CAS 33156-15-9
Molecular Formula C15H21N5O4
Molecular Weight (g/mol) 335.36
MDL Number MFCD00005741
InChI Key USVMJSALORZVDV-SDBHATRESA-N
Synonym 2r,3s,4r,5r-2-hydroxymethyl-5-6-3-methylbut-2-enylamino purin-9-yl oxolane-3,4-diol, n-6-delta-2-isopentenyl adenosine hemihydrate, 99+%
PubChem CID 133126474
IUPAC Name (2R,3S,4R,5R)-2-(hydroxymethyl)-5-{6-[(3-methylbut-2-en-1-yl)amino]-9H-purin-9-yl}oxolane-3,4-diol
SMILES CC(C)=CCNC1=C2N=CN([C@@H]3O[C@H](CO)[C@@H](O)[C@H]3O)C2=NC=N1

Specifications

CAS Min % 99.0
CAS Max % 100.0
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 99+%
Quantity 100 mg
Formula Weight 335.35
Percent Purity ≥99.0%
Physical Form Crystalline Powder
Chemical Name or Material N-6-(δ-2-Isopentenyl)adenosine hemihydrate, 99+%

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.