Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Acetyl-D-Glucosamine Kinase/NAGK Antibody, PE, Novus Biologicals™
SDP

Catalog No. NB291573 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB291573 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB291573 Supplier Novus Biologicals Supplier No. NBP142694PE
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

N-Acetyl-D-Glucosamine Kinase/NAGK Polyclonal antibody specifically detects N-Acetyl-D-Glucosamine Kinase/NAGK in Human samples. It is validated for Western Blot, Immunoprecipitation

Specifications

Antigen N-Acetyl-D-Glucosamine Kinase/NAGK
Applications Western Blot, Immunoprecipitation
Classification Polyclonal
Conjugate PE
Dilution Western Blot, Immunoprecipitation
Formulation PBS
Gene Alias EC 2.7.1.59, GNKGlcNAc kinase, N-acetyl-D-glucosamine kinase, N-acetylglucosamine kinaseHSA242910
Host Species Rabbit
Immunogen The immunogen for this product maps to a region between residue 50 and 100 of human N-acetylglucosamine kinase using the numbering given in entry NP_060037.2 (GeneID 55577).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline metabolism
Primary or Secondary Primary
Gene ID (Entrez) 55577
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.