Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Acetyl-L-methionine, 99%

Catalog No. p-7027509
Click to view available options
Quantity:
5 g
25 g

CAS: 65-82-7 | C7H13NO3S | 191.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 65-82-7
Molecular Formula C7H13NO3S
Molecular Weight (g/mol) 191.25
MDL Number MFCD00064441
InChI Key XUYPXLNMDZIRQH-LURJTMIESA-N
Synonym n-acetyl-l-methionine, n-acetylmethionine, acetyl-l-methionine, ac-met-oh, l-n-acetyl methionine, acetylmethionine, methionamine, methionine, n-acetyl-, l, thiomedon, l-methionine, n-acetyl
PubChem CID 448580
ChEBI CHEBI:21557
IUPAC Name (2S)-2-acetamido-4-methylsulfanylbutanoic acid
SMILES CSCC[C@H](NC(C)=O)C(O)=O

Specifications

Melting Point 103°C to 107°C
Beilstein 1725552
Merck Index 14,96
Quantity 5 g
Formula Weight 191.25
Optical Rotation −21° (c=1.3 in Water)
Percent Purity 99%
Chemical Name or Material N-Acetyl-L-methionine
EINECSNumber 200-617-7
RTECSNumber PD0480000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.