Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Acetylneuraminic Acid Hydrate 98.0+%, TCI America™
SDP

Catalog No. p-7116981
Click to view available options
Quantity:
100 mg
1 g
5 g

Chemical Identifiers

CAS 131-48-6
Molecular Formula C11H19NO9
Molecular Weight (g/mol) 309.271
MDL Number MFCD00006620
InChI Key SQVRNKJHWKZAKO-LFIUDZTESA-N
Synonym n-acetylneuramic acid, spectrum_000450, spectrum2_000559, spectrum4_000432, spectrum5_001326, spectrum2300147, 2s,4s,5s,6s-5-acetamido-2,4-dihydroxy-6-2r-1,2,3-trihydroxypropyl oxane-2-carboxylic acid
PubChem CID 126963458
IUPAC Name (4S,5S,6R)-5-acetamido-2,4-dihydroxy-6-[(1R,2R)-1,2,3-trihydroxypropyl]oxane-2-carboxylic acid
SMILES CC(=O)NC1C(CC(OC1C(C(CO)O)O)(C(=O)O)O)O

Specifications

Melting Point 186°C
Color White-Yellow
Quantity 100 mg
Formula Weight 309.27
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material N-Acetylneuraminic Acid Hydrate
EINECSNumber (5)-6979
TSCA Yes
Recommended Storage Freezer
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.