Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
N-acetylprocainamide Monoclonal Antibody (NAPA11-128.2.1), QED Bioscience™

Mouse Monoclonal Antibody
$250.22 - $1002.00
Specifications
Antigen | N-acetylprocainamide |
---|---|
Clone | NAPA11-128.2.1 |
Concentration | 0.5-1.0 mg/mL |
Applications | ELISA |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-227-7780
![]() ![]() |
Leinco Technologies Inc
16908100UG |
100 μg |
Each for $250.22
|
|
|||||
50-227-7781
![]() ![]() |
Leinco Technologies Inc
16908500UG |
500 μg |
Each for $500.43
|
|
|||||
50-227-7779
![]() ![]() |
Leinco Technologies Inc
169081000UG |
1 mg |
Each for $1,002.00
|
|
|||||
Description
N-acetylprocainamide Monoclonal antibody specifically detects N-acetylprocainamide in Chemical samples. It is validated for ELISASpecifications
N-acetylprocainamide | |
0.5-1.0 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
Acecainida; Acecainide; Acecainidum; Acekainid; C15H23N3O2; N-Acetyloprokainamid | |
IgG1 | |
Protein A | |
Antibody |
NAPA11-128.2.1 | |
ELISA | |
Unconjugated | |
Mouse | |
PBS with no preservative; pH 7.4 | |
N-Acetylprocainamide-KLH. | |
Primary | |
-20° C, Avoid Freeze/Thaw Cycles |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title