Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-acylmannosamine 1-dehydrogenase Antibody, Biotin, Novus Biologicals™
SDP

Catalog No. NBP17770801 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP17770801 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP17770801 Supplier Novus Biologicals Supplier No. NBP1777080.1MG
Add to Cart
Add to Cart

Goat Polyclonal Antibody

N-acylmannosamine 1-dehydrogenase Polyclonal specifically detects N-acylmannosamine 1-dehydrogenase in Bacteria samples. It is validated for Western Blot, ELISA, Immunoprecipitation.

Specifications

Antigen N-acylmannosamine 1-dehydrogenase
Applications Western Blot, ELISA, Immunoprecipitation
Classification Polyclonal
Conjugate Biotin
Dilution Western Blot 1:500 - 1:5000, ELISA 1:5000 - 1:20000, Immunoprecipitation 1:100
Formulation 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2, 10 mg/mL Bovine Serum Albumin (BSA) - Immunoglobulin and Protease free with 0.01% Sodium Azide
Host Species Goat
Immunogen N-Acyl Mannosamine-1-Dehydrogenase [Recombinant Sequence from Pseudomonas expressed in E.coli ]
Purification Method Delipidation and Defibrination
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Bacteria
Content And Storage Store at 4C. Do not freeze.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.