Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Cadherin Antibody (001), DyLight 350
SDP

Catalog No. NB067845 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB067845 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB067845 Supplier Novus Biologicals Supplier No. NBP289752UV

Rabbit Monoclonal Antibody

N-Cadherin Monoclonal specifically detects N-Cadherin in Human samples. It is validated for Flow Cytometry, ELISA.

Specifications

Antigen N-Cadherin
Applications Flow Cytometry, ELISA
Classification Monoclonal
Clone 1
Conjugate DyLight 350
Dilution Flow Cytometry, ELISA
Formulation 50mM Sodium Borate
Gene Alias cadherin 2, N-cadherin (neuronal), cadherin 2, type 1, N-cadherin (neuronal), cadherin-2, CD325, CD325 antigen, CDHNcalcium-dependent adhesion protein, neuronal, CDw325, N-cadherin, NCADN-cadherin 1, Neural cadherin, neural-cadherin
Host Species Rabbit
Immunogen This Recombinant N-Cadherin Antibody (001) was obtained from a rabbit immunized with purified, recombinant Human N-Cadherin (NP_001783.2; Met 1-Ala 724).
Molecular Weight of Antigen 100 kDa
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cell Cycle and Replication, Cellular Markers, Extracellular Matrix, Mesenchymal Stem Cell Markers, Neuroscience, Plasma Membrane Markers, Signal Transduction, Stem Cells
Primary or Secondary Primary
Gene ID (Entrez) 1000
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.