Learn More
Description
Specifications
Specifications
| Antigen | N-Cadherin |
| Applications | ELISA, Immunofluorescence |
| Classification | Monoclonal |
| Clone | 14 |
| Conjugate | Biotin |
| Formulation | PBS |
| Gene Alias | cadherin 2, N-cadherin (neuronal), cadherin 2, type 1, N-cadherin (neuronal), cadherin-2, CD325, CD325 antigen, CDHNcalcium-dependent adhesion protein, neuronal, CDw325, N-cadherin, NCADN-cadherin 1, Neural cadherin, neural-cadherin |
| Host Species | Rabbit |
| Immunogen | This Recombinant N-Cadherin Antibody (014) was obtained from a rabbit immunized with purified, recombinant Human N-Cadherin (NP_001783.2; Met 1-Ala 724). |
| Molecular Weight of Antigen | 100 kDa |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
