Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Cadherin Antibody (GC-4), Novus Biologicals™
SDP

Catalog No. NBP22180501 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.05 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP22180501 0.1 mL
NBP22180505 0.05 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP22180501 Supplier Novus Biologicals Supplier No. NBP2218050.1ML
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

N-Cadherin Monoclonal specifically detects N-Cadherin in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen, In vitro assay, Electron Microscopy.

Specifications

Antigen N-Cadherin
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen)
Classification Monoclonal
Clone GC-4
Conjugate Unconjugated
Dilution Western Blot 10-20μ/mL, Flow Cytometry 1:50-1:250, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:50-1:250, Immunohistochemistry-Frozen 10-20μ/mL, Electron Microscopy, Immunofluorescence
Formulation 10 mM PBS (pH 7.4) with no preservative
Gene Alias cadherin 2, N-cadherin (neuronal), cadherin 2, type 1, N-cadherin (neuronal), cadherin-2, CD325, CD325 antigen, CDHNcalcium-dependent adhesion protein, neuronal, CDw325, N-cadherin, NCADN-cadherin 1, Neural cadherin, neural-cadherin
Gene Symbols CDH2
Host Species Mouse
Immunogen Affinity purified chicken heart A-CAM.
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 1000
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.