Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
N-Cadherin Antibody (GC-4), Novus Biologicals™

Mouse Monoclonal Antibody
$316.50 - $611.00
Specifications
Antigen | N-Cadherin |
---|---|
Clone | GC-4 |
Dilution | Western Blot 10-20μ/mL, Flow Cytometry 1:50-1:250, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:50-1:250, Immunohistochemistry-Frozen 10-20μ/mL, Electron Microscopy, Immunofluorescence |
Applications | Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen) |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP22180505
![]() |
Novus Biologicals
NBP2218050.05ML |
0.05 mL |
Each for $316.50
|
|
|||||
NBP22180501
![]() |
Novus Biologicals
NBP2218050.1ML |
0.1 mL |
Each for $611.00
|
|
|||||
Description
N-Cadherin Monoclonal specifically detects N-Cadherin in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen, In vitro assay, Electron Microscopy.Specifications
N-Cadherin | |
Western Blot 10-20μ/mL, Flow Cytometry 1:50-1:250, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:50-1:250, Immunohistochemistry-Frozen 10-20μ/mL, Electron Microscopy, Immunofluorescence | |
Monoclonal | |
Mouse | |
10 mM PBS (pH 7.4) with no preservative | |
1000 | |
Affinity purified chicken heart A-CAM. | |
Primary |
GC-4 | |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen) | |
Unconjugated | |
Human, Mouse, Rat | |
cadherin 2, N-cadherin (neuronal), cadherin 2, type 1, N-cadherin (neuronal), cadherin-2, CD325, CD325 antigen, CDHNcalcium-dependent adhesion protein, neuronal, CDw325, N-cadherin, NCADN-cadherin 1, Neural cadherin, neural-cadherin | |
CDH2 | |
IgG1 | |
Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title