Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Cadherin Antibody (GC-4), Novus Biologicals™
SDP

Catalog No. NBP221805 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.2 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP221805 0.2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP221805 Supplier Novus Biologicals Supplier No. NBP221805
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

N-Cadherin Monoclonal specifically detects N-Cadherin in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen, In vitro assay, Electron Microscopy.

Specifications

Antigen N-Cadherin
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen), In vitro Assay, Immunoassay
Classification Monoclonal
Clone GC-4
Conjugate Unconjugated
Dilution Western Blot 10-20ug/mL, Flow Cytometry 1:50-1:250, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:50-1:250, Immunohistochemistry-Frozen 10-20ug/mL, In vitro assay, Electron Microscopy
Gene Alias cadherin 2, N-cadherin (neuronal), cadherin 2, type 1, N-cadherin (neuronal), cadherin-2, CD325, CD325 antigen, CDHNcalcium-dependent adhesion protein, neuronal, CDw325, N-cadherin, NCADN-cadherin 1, Neural cadherin, neural-cadherin
Gene Symbols CDH2
Host Species Mouse
Immunogen This N-Cadherin Antibody (GC-4) was developed against affinity purified chicken heart A-CAM.
Molecular Weight of Antigen 135 kDa
Purification Method Protein A or G purified
Quantity 0.2 mL
Regulatory Status RUO
Research Discipline Cancer, Cell Cycle and Replication, Cellular Markers, Extracellular Matrix, Mesenchymal Stem Cell Markers, Neuroscience, Plasma Membrane Markers, Signal Transduction, Stem Cells
Primary or Secondary Primary
Gene ID (Entrez) 1000
Test Specificity Chicken, Monkey, Rabbit, Human, Mouse, Rat.
Target Species Human, Mouse, Rat
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.