Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Cadherin Antibody (OTI2G7), PE/Cy7, Novus Biologicals™
SDP

Catalog No. N272891PEC7 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N272891PEC7 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N272891PEC7 Supplier Novus Biologicals Supplier No. NBP272891PECY7
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

N-Cadherin Monoclonal specifically detects N-Cadherin in Human, Mouse, Rat, Canine, Monkey, Primate samples. It is validated for Flow Cytometry.

Specifications

Antigen N-Cadherin
Applications Flow Cytometry
Classification Monoclonal
Clone OTI2G7
Conjugate PE-Cyanine7
Dilution Flow Cytometry
Formulation PBS with 0.05% Sodium Azide
Gene Alias cadherin 2, N-cadherin (neuronal), cadherin 2, type 1, N-cadherin (neuronal), cadherin-2, CD325, CD325 antigen, CDHNcalcium-dependent adhesion protein, neuronal, CDw325, N-cadherin, NCADN-cadherin 1, Neural cadherin, neural-cadherin
Gene Symbols CDH2
Host Species Mouse
Immunogen This N-Cadherin Antibody (OTI2G7) [PE/Cy7] was developed against a full length recombinant protein of human N-Cadherin (NP_001783) produced in HEK293T cells.
Molecular Weight of Antigen 97.2 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Cancer, Cell Cycle and Replication, Cellular Markers, Extracellular Matrix, Mesenchymal Stem Cell Markers, Neuroscience, Plasma Membrane Markers, Signal Transduction, Stem Cells
Primary or Secondary Primary
Gene ID (Entrez) 1000.0
Target Species Human, Mouse, Rat, Canine, Monkey, Primate
Content And Storage Store at 4°C in the dark. Do not freeze.
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.