Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-ε-Carbobenzyloxy-L-lysine, 98%, Thermo Scientific Chemicals

Catalog No. ac108360010
Click to view available options
Quantity:
1 g
25 g
Packaging:
Glass Bottle

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1155-64-2
Molecular Formula C14H20N2O4
Molecular Weight (g/mol) 280.32
MDL Number MFCD00002638
InChI Key CKGCFBNYQJDIGS-LBPRGKRZSA-N
Synonym h-lys z-oh, n6-cbz-l-lysine, n-epsilon-carbobenzyloxy-l-lysine, n epsilon-benzyloxycarbonyl-l-lysine, n6-benzyloxycarbonyl-l-lysine, nepsilon-carbobenzoxy-l-lysine, nepsilon-carbobenzyloxy-l-lysine, l-lys cbz-oh, 2s-2-amino-6-benzyloxy carbonyl amino hexanoic acid, 6-n-cbz-l-lysine
PubChem CID 1715626
SMILES N[C@@H](CCCCNC(=O)OCC1=CC=CC=C1)C(O)=O

Specifications

Melting Point 259.0°C
CAS Min % 97.5
CAS Max % 100.0
Color White
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass Bottle
Linear Formula C6H5CH2O2CNH(CH2)4CH(NH2)CO2H
Specific Rotation + 15.50
Sulfated Ash 0.2% max.
Solubility Information Solubility in water: .. Other solubilities: soluble in most common organic solvents
Water 0.5% max. (K.F.)
IUPAC Name (2S)-2-amino-6-{[(benzyloxy)carbonyl]amino}hexanoic acid
Specific Rotation Condition +15.50 (20.00°C c=1,1N HCl)
Quantity 1 g
Formula Weight 280.32
Percent Purity 98%
Physical Form Powder
Chemical Name or Material Nε-Carbobenzyloxy-L-lysine
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.