Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Deacetylase/N-Sulfotransferase 1/NDST1 Antibody, mFluor Violet 610 SE, Novus Biologicals™
SDP

Catalog No. NB342513 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB342513 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB342513 Supplier Novus Biologicals Supplier No. NBP281758MFV610
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

N-Deacetylase/N-Sulfotransferase 1/NDST1 Polyclonal antibody specifically detects N-Deacetylase/N-Sulfotransferase 1/NDST1 in Human samples. It is validated for Western Blot, ELISA

Specifications

Antigen N-Deacetylase/N-Sulfotransferase 1/NDST1
Applications Western Blot, ELISA
Classification Polyclonal
Conjugate mFluor Violet 610 SE
Dilution Western Blot, ELISA
Formulation 50mM Sodium Borate
Gene Alias [Heparan sulfate]-glucosamine N-sulfotransferase 1, bifunctional heparan sulfate N-deacetylase/N-sulfotransferase 1, EC 2.8.2.8, Glucosaminyl N-deacetylase/N-sulfotransferase 1, heparan sulfate-N-deacetylase/N-sulfotransferase, HSNST 1, HSST1, HSSTN-HSST 1, N-deacetylase/N-sulfotransferase (heparan glucosaminyl) 1, NDST-1, N-heparan sulfate sulfotransferase 1, NST1
Host Species Rabbit
Immunogen NDST1 antibody was raised against a peptide corresponding to 14 amino acids near the amino terminus of human NDST1. Amino Acid Squence: SRFKYRTEIAPGKGDMPT
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 3340
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.