Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Deacetylase/N-Sulfotransferase 2/NDST2 Antibody, DyLight 488, Novus Biologicals™
SDP

Catalog No. NB164586 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB164586 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB164586 Supplier Novus Biologicals Supplier No. NBP281747G
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

N-Deacetylase/N-Sulfotransferase 2/NDST2 Polyclonal antibody specifically detects N-Deacetylase/N-Sulfotransferase 2/NDST2 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunofluorescence

Specifications

Antigen N-Deacetylase/N-Sulfotransferase 2/NDST2
Applications Western Blot, ELISA, Immunofluorescence
Classification Polyclonal
Conjugate DyLight 488
Formulation 50mM Sodium Borate
Gene Alias bifunctional heparan sulfate N-deacetylase/N-sulfotransferase 2, EC 2.8.2.8, Glucosaminyl N-deacetylase/N-sulfotransferase 2, HSST2MGC129697, MGC129705, N-deacetylase/N-sulfotransferase (heparan glucosaminyl) 2, NDST-2, N-heparan sulfate sulfotransferase 2, N-HSST 2, NST2
Host Species Rabbit
Immunogen NDST2 antibody was raised against a 16 amino acid peptide near the amino terminus of human NDST2. The immunogen is located within amino acids 20 - 70 of NDST2. Amino Acid Squence: YVSTSPKAKEPLPLPL
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8509
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.