Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

n-Decane, 99+%

Catalog No. p-7049919
Click to view available options
Quantity:
10 g
100 g
500 g

CAS: 124-18-5 | C10H22 | 142.286 g/mol

n-Decane is used as a solvent as well as an internal standard in the GC analysis of oxalic, malonic and succinic acids. It is used in paint manufacture as a hydrocarbon solvent and for wood stains and varnishes as a varnish solvent. It is an active component of automotive fuels viz. gasoline (petrol).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 124-18-5
Molecular Formula C10H22
Molecular Weight (g/mol) 142.286
MDL Number MFCD00008954
InChI Key DIOQZVSQGTUSAI-UHFFFAOYSA-N
Synonym n-decane, nonane, methyl, decyl hydride, unii-nk85062oiy, ccris 653, hsdb 63, dsstox_cid_4913, decane, analytical standard, dsstox_rid_77577, dsstox_gsid_24913
PubChem CID 15600
ChEBI CHEBI:41808
IUPAC Name decane
SMILES CCCCCCCCCC

Specifications

Melting Point -30°C
Color Colorless
Density 0.730 g/mL
Boiling Point 172°C to 174°C
Flash Point 46°C (115°F)
Odor Petrol-like
Assay Percent Range >99%
Refractive Index 1.4115
Linear Formula CH3(CH2)8CH3
Quantity 10 g
UN Number UN2247
Beilstein 1696981
Solubility Information Miscible with ethanol and ether. Slightly miscible with carbon tetrachloride. Immiscible with water.
Formula Weight 142.29
Percent Purity ≥99%
Physical Form Liquid
Chemical Name or Material n-Decane
Show More Show Less
Hazard Category H226-H304
Hazard Statement GHS H Statement
H304-H226
May be fatal if swallowed and enters airways.
Flammable liquid and vapor.
Precautionary Statement P210-P233-P240-P241-P242-P243-P280-P301+P310-P303+P361+P353-P331-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: N-DECANE
EINECSNumber 204-686-4
RTECSNumber HD6550000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.