Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

n-Decane, 99%, Spectrum™ Chemical
SDP

Catalog No. 18611918 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
13 kg
150 kg
Packaging:
Amber Glass Bottle
Drum
Steel Pail
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-611-918 13 kg Steel Pail
18-611-917 100 mL Amber Glass Bottle
18-611-920 500 mL Amber Glass Bottle
18-611-919 150 kg Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18-611-918 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. D209813KGBL
Add to Cart
Add to Cart

Chemical Identifiers

CAS 124-18-5

Specifications

CAS Max % 1
Packaging Steel Pail
Quantity 13 kg
Percent Purity 0.99
Grade Reagent

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.