Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Decyl-N,N-dimethyl-3-ammonio-1-propanesulfonate, 98%

Catalog No. AC459680250
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC459680250 25 g
AC459680050 5 g
AC459681000 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC459680250 Supplier Thermo Scientific Chemicals Supplier No. 459680250
Add to Cart

CAS: 15163-36-7 | C15H33NO3S | 307.49 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15163-36-7
Molecular Formula C15H33NO3S
Molecular Weight (g/mol) 307.49
MDL Number MFCD00036908
InChI Key WKALLSVICJPZTM-UHFFFAOYSA-N
Synonym 3-decyldimethylammonio propane-1-sulfonate, n-decyl-n,n-dimethyl-3-ammonio-1-propanesulfonate, sulfobetaine 10, caprylyl sulfobetaine, 3-decyldimethylammonio propanesulfonate, zwittergent 3-10 detergent, capryl sultaine, zwittergent 310, 3-decyl dimethyl ammonio propane-1-sulfonate, 3-decyldimethylamino propanesulfonic acid
PubChem CID 161111
IUPAC Name 3-[decyl(dimethyl)azaniumyl]propane-1-sulfonate
SMILES CCCCCCCCCC[N+](C)(C)CCCS([O-])(=O)=O

Specifications

Melting Point 229.0°C
Loss on Drying 2% max.
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Quantity 25 g
Solubility Information (10% in water) Clear colorless solution
Formula Weight 307.5
Percent Purity 98%
Chemical Name or Material N-Decyl-N,N-dimethyl-3-ammonio-1-propanesulfonate
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective g

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.