Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-(Diphenylmethylene)aminoacetonitrile, 99%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
70591-20-7
C15H12N2
220.27
MFCD00009970
VRLJFRODHVSTIK-UHFFFAOYSA-N
n-diphenylmethylene aminoacetonitrile,2-diphenylmethylene amino acetonitrile,benzhydrylidene-amino-acetonitrile,2-benzhydrylideneamino acetonitrile,2-diphenylmethylidene amino acetonitrile,acetonitrile, diphenylmethylene amino,4,4-diphenyl-3-azabut-3-enenitrile,pubchem13883,benzhydrylideneaminoacetonitrile
612957
2-(benzhydrylideneamino)acetonitrile
C1=CC=C(C=C1)C(=NCC#N)C2=CC=CC=C2
70591-20-7
C15H12N2
220.27
MFCD00009970
VRLJFRODHVSTIK-UHFFFAOYSA-N
n-diphenylmethylene aminoacetonitrile,2-diphenylmethylene amino acetonitrile,benzhydrylidene-amino-acetonitrile,2-benzhydrylideneamino acetonitrile,2-diphenylmethylidene amino acetonitrile,acetonitrile, diphenylmethylene amino,4,4-diphenyl-3-azabut-3-enenitrile,pubchem13883,benzhydrylideneaminoacetonitrile
612957
2-(benzhydrylideneamino)acetonitrile
C1=CC=C(C=C1)C(=NCC#N)C2=CC=CC=C2
Melting Point 83.0°C to 86.0°C
Infrared Spectrum Authentic
Color Gray or White to Yellow
Packaging Glass bottle
Quantity 5g
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Linear Formula (C6H5)2C=NCH2CN
Formula Weight 220.27
Physical Form Powder or Crystals
Percent Purity 99%
Chemical Name or Material N-(Diphenylmethylene)aminoacetonitrile
Show More Show Less