Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

n-Dodecane, 99%

Catalog No. AA3132414
Click to view available options
Quantity:
25 g
100 mL
500 g
500 mL
2500 mL

CAS: 112-40-3 | C12H26 | 170.34 g/mol

n-Dodecane is used as a solvent and a distillation chaser. It finds application as a diluent for tributyl phosphate (TBP) in reprocessing plants and as a possible surrogate for kerosene-based fuels in jet. It is an active component of scintillator as well as used in lubricants and greases.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
n-Dodecane is used as a solvent and a distillation chaser. It finds application as a diluent for tributyl phosphate (TBP) in reprocessing plants and as a possible surrogate for kerosene-based fuels in jet. It is an active component of scintillator as well as used in lubricants and greases.

Solubility
Miscible with ethyl alcohol, ethyl ether, acetone, chloroform and carbon tetrachloride. Immiscible with water.

Notes
Hygroscopic. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 112-40-3
Molecular Formula C12H26
Molecular Weight (g/mol) 170.34
MDL Number MFCD00008969
InChI Key SNRUBQQJIBEYMU-UHFFFAOYSA-N
Synonym n-dodecane, dihexyl, bihexyl, adakane 12, n-dodecan, n-dodecan german, duodecane, alkanes, c10-14, undecane, methyl, ccris 661
PubChem CID 8182
ChEBI CHEBI:28817
IUPAC Name dodecane
SMILES CCCCCCCCCCCC

Specifications

Melting Point -10°C
Color Colorless
Density 0.749 g/mL
Boiling Point 215°C to 217°C
Flash Point 71°C (159°F)
Odor Petrol-like
Refractive Index 1.422
Linear Formula CH3(CH2)10CH3
Quantity 25 g
Beilstein 1697175
Solubility Information Miscible with ethyl alcohol,ethyl ether,acetone,chloroform and carbon tetrachloride. Immiscible with water.
Formula Weight 170.34
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material n-Dodecane
Show More Show Less
Hazard Category H227-H304
Hazard Statement GHS H Statement
H304-H227
May be fatal if swallowed and enters airways.
Combustible liquid.
Precautionary Statement P210-P280-P301+P310-P331-P370+P378q-P501c
EINECSNumber 203-967-9
RTECSNumber JR2125000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.