Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Ethyl-2-nitroaniline, 98%, Thermo Scientific™

Catalog No. AAH55148MD
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH55148MD 250 mg
AAH5514803 1 g
AAH5514806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH55148MD Supplier Thermo Scientific Chemicals Supplier No. H55148MD

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10112-15-9
Molecular Formula C8H10N2O2
Molecular Weight (g/mol) 166.18
MDL Number MFCD00007091
InChI Key CQIKVOWCSGXCCG-UHFFFAOYSA-N
Synonym benzenamine,n-ethyl-2-nitro, benzenamine, n-ethyl-2-nitro, n-ethyl-2-nitroaniline, ethyl-2-nitro-phenyl-amine, ethyl-6-nitroaniline, acmc-20ak4v, ethyl 2-nitrophenyl amine, n-ethyl-n-2-nitrophenyl amine #
PubChem CID 82354
IUPAC Name N-ethyl-2-nitroaniline
SMILES CCNC1=CC=CC=C1[N+](=O)[O-]

Specifications

Color Colorless to Orange
Density 1.19 g/mL
Boiling Point 168°C (20 mmHg)
Flash Point 113°C (235°F)
Refractive Index 1.645
Quantity 250 mg
Formula Weight 166.18
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material N-Ethyl-2-nitroaniline
Hazard Category H302+H312+H332-H315-H319-H335
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 233-305-4
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.