Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Ethyl-p-menthane-3-carboxamide 98.0+%, TCI America™
SDP

Catalog No. E079625G Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
5 g
25 g

Catalog No. E079625G

Supplier: TCI America E079625G

Only null left
Add to Cart
Add to Cart

Chemical Identifiers

CAS 39711-79-0
Molecular Formula C13H25NO
Molecular Weight (g/mol) 211.35
MDL Number MFCD00130071
InChI Key VUNOFAIHSALQQH-UHFFFAOYNA-N
Synonym n-ethyl-p-menthane-3-carboxamide, ethyl menthane carboxamide, n-ethyl-2-isopropyl-5-methylcyclohexanecarboxamide, ws-3, n-ethyl-4-menthane-3-carboxamide, cyclohexanecarboxamide, n-ethyl-5-methyl-2-1-methylethyl, fema no. 3455, ws 3, n-ethyl-2-isopropyl-5-methylcyclohexane carboxamide
PubChem CID 62907
IUPAC Name N-ethyl-5-methyl-2-(propan-2-yl)cyclohexane-1-carboxamide
SMILES CCNC(=O)C1CC(C)CCC1C(C)C

Specifications

Melting Point 99°C
Color White
Quantity 25 g
Formula Weight 211.35
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material N-Ethyl-p-menthane-3-carboxamide
EINECSNumber (3)-4186
TSCA Yes
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.