Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ N-Fmoc-2-methyl-D-phenylalanine, 95%

Click to view available options
Quantity:
1g
5g
352351-63-4
C25H23NO4
401.462
MFCD00671404
GYFMRMRGTNDDAT-HSZRJFAPSA-N
fmoc-2-methyl-d-phenylalanine,r-2-9h-fluoren-9-yl methoxy carbonyl amino-3-o-tolyl propanoic acid,fmoc-d-2-methylphenylalanine,fmoc-d-2-methylphe,fmoc-d-phe 2-me-oh,fmoc-d-2-me-phe-oh,fmoc-l-phe 2-me-oh,2r-2-9h-fluoren-9-ylmethoxy carbonyl amino-3-2-methylphenyl propanoic acid,fmoc-o-me-d-phe-oh,fmoc-2-methy-d-phenylalanine
7129821
(2R)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-(2-methylphenyl)propanoic acid
CC1=CC=CC=C1CC(C(=O)O)NC(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24
352351-63-4
C25H23NO4
401.462
MFCD00671404
GYFMRMRGTNDDAT-HSZRJFAPSA-N
fmoc-2-methyl-d-phenylalanine,r-2-9h-fluoren-9-yl methoxy carbonyl amino-3-o-tolyl propanoic acid,fmoc-d-2-methylphenylalanine,fmoc-d-2-methylphe,fmoc-d-phe 2-me-oh,fmoc-d-2-me-phe-oh,fmoc-l-phe 2-me-oh,2r-2-9h-fluoren-9-ylmethoxy carbonyl amino-3-2-methylphenyl propanoic acid,fmoc-o-me-d-phe-oh,fmoc-2-methy-d-phenylalanine
7129821
(2R)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-(2-methylphenyl)propanoic acid
CC1=CC=CC=C1CC(C(=O)O)NC(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24
Beilstein 9157614
Formula Weight 401.46
Physical Form Powder
Percent Purity 95%
Color White
Melting Point 110°C to 116°C
Quantity 1g
Chemical Name or Material N-Fmoc-2-methyl-D-phenylalanine

storageNote1 : 2° to 8°C