Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Fmoc-3-(2-naphthyl)-D-alanine, 95%

Catalog No. p-7034303
Click to view available options
Quantity:
1 g
5 g

CAS: 138774-94-4 | C28H23NO4 | 437.50 g/mol

N-Fmoc-3-(2-naphthyl)-D-alanine is an Fmoc-protected alanine derivative and an intermediate used in the preparation of phosphoserine-containing tetrapeptides with hydrophobic N-terminal acyl groups as inhibitors of the BRCA1 protein.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Fmoc-3-(2-naphthyl)-D-alanine is an Fmoc-protected alanine derivative and an intermediate used in the preparation of phosphoserine-containing tetrapeptides with hydrophobic N-terminal acyl groups as inhibitors of the BRCA1 protein.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 138774-94-4
Molecular Formula C28H23NO4
Molecular Weight (g/mol) 437.50
MDL Number MFCD00077044
InChI Key JYUTZJVERLGMQZ-AREMUKBSSA-N
Synonym fmoc-d-2-nal-oh, fmoc-3-2-naphthyl-d-alanine, r-2-9h-fluoren-9-yl methoxy carbonyl amino-3-naphthalen-2-yl propanoic acid, fmoc-d-3-2-naphthyl-alanine, n-fmoc-3-2-naphthyl-d-alanine, fmoc-d-2-naphthylalanine, s-n-fmoc-2-naphthylalanine, fmoc-beta-2-naphthyl-d-alanine, 2r-2-9h-fluoren-9-ylmethoxy carbonyl amino-3-naphthalen-2-yl propanoic acid, pubchem11948
PubChem CID 7130590
IUPAC Name (2R)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-naphthalen-2-ylpropanoic acid
SMILES OC(=O)[C@@H](CC1=CC2=CC=CC=C2C=C1)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12

Specifications

Quantity 1 g
Beilstein 7864489
Solubility Information Slightly soluble in water.
Formula Weight 437.49
Percent Purity 95%
Chemical Name or Material N-Fmoc-3-(2-naphthyl)-D-alanine
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.