Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Fmoc-S-ethyl-L-cysteine, 95%, Thermo Scientific™

Catalog No. AAH6601503
Click to view available options
Quantity:
250 mg
1 g
5 g

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 200354-34-3
Molecular Formula C20H21NO4S
Molecular Weight (g/mol) 371.451
MDL Number MFCD00151921
InChI Key OUXUHWHQQLGDQE-SFHVURJKSA-N
Synonym fmoc-cys et-oh, fmoc-s-ethyl-l-cysteine, l-cysteine,s-ethyl-n-9h-fluoren-9-ylmethoxy carbonyl, 2r-3-ethylsulfanyl-2-9h-fluoren-9-ylmethoxy carbonyl amino propanoic acid, s-ethyl-n-9h-fluoren-9-yl methoxy carbonyl-l-cysteine
PubChem CID 7015833
IUPAC Name (2R)-3-ethylsulfanyl-2-(9H-fluoren-9-ylmethoxycarbonylamino)propanoic acid
SMILES CCSCC(C(=O)O)NC(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13

Specifications

Quantity 1 g
Formula Weight 371.45
Percent Purity 95%
Physical Form Solid
Chemical Name or Material N-Fmoc-S-ethyl-L-cysteine
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
storageNote1 -30° to -10°C
Recommended Storage Store at -20°C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.