Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
n-Heptane, HPLC Grade, J.T. Baker™
Shop All Avantor Products
Click to view available options
Quantity:
4 L
Packaging:
Narrow Mouth Amber Glass Bottle
Narrow Mouth PVC-coated Amber Glass Bottle
Description
- For use in liquid chromatography (HPLC & UHPLC) & spectrophotometry; For laboratory, research, or manufacturing use
Specifications
Specifications
| Quantity | 4 L |
| Flash Point | −4.00°C (−39.2.00°F) |
| Packaging | Narrow Mouth Amber Glass Bottle |
| Specific Gravity | 1L = 0.68kg |
| Formula Weight | 100.2 |
| Grade | Meets ACS reagent chemical requirements |
| Chemical Name or Material | n-Heptane |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction