Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

n-Hexadecane, 99.5+%

Catalog No. p-7059376
Click to view available options
Quantity:
5 g
25 g

CAS: 544-76-3 | C16H34 | 226.448 g/mol

Employed in industry as Adsorbents, Fuels and fuel additives. Also in Fabric, Textile, and Leather Products. Solvent, organic intermediate, ignition standard for diesel fuels. Fuels and Related Products. Hexadecane is use in the recovery of valuable amines by means of distillation from aqueous alkanolamine solutions, which are extensively used in the gas-processing industries.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 544-76-3
Molecular Formula C16H34
Molecular Weight (g/mol) 226.448
MDL Number MFCD00008998
InChI Key DCAYPVUWAIABOU-UHFFFAOYSA-N
Synonym n-hexadecane, cetane, n-cetane, hexadecan, hexadekan, cetan, zetan, unii-f8z00shp6q, ccris 5833, pentadecane, methyl
PubChem CID 11006
ChEBI CHEBI:45296
IUPAC Name hexadecane
SMILES CCCCCCCCCCCCCCCC

Specifications

Melting Point 18°C
Color Colorless
Density 0.773 g/mL
Boiling Point 287°C
Flash Point 135°C (275°F)
Refractive Index 1.4345
Quantity 5 g
Beilstein 1736592
Solubility Information Soluble in alcohol,acetone,ether.
Formula Weight 226.45
Percent Purity ≥99.5%
Physical Form Liquid
Chemical Name or Material n-Hexadecane
Show More Show Less
Hazard Category H304
Hazard Statement GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P301+P310-P331-P501c
EINECSNumber 208-878-9
RTECSNumber ML9200000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.