Learn More
n-Hexatriacontane, 97+%
Description
n-Hexatriacontane is used as a paraffin wax and in candles. It is used as a key starting material for the preparation of lower alkanes by cracking. It is also used for example in road surfacing.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Melting Point | 76.2°C |
Color | White |
Density | 0.779 g/mL |
Boiling Point | 497°C |
Refractive Index | 1.4573 |
Quantity | 50 g |
Solubility Information | Insoluble in water. |
Formula Weight | 506.95 |
Percent Purity | ≥97% |
Physical Form | Wax |
Show More |
Safety and Handling
EINECSNumber | 211-127-8 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.