Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Isopropylbenzylamine, 97%

Catalog No. aab2324322
Click to view available options
Quantity:
100 g
500 g

CAS: 102-97-6 | C10H15N | 149.237 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 102-97-6
Molecular Formula C10H15N
Molecular Weight (g/mol) 149.237
MDL Number MFCD00008863
InChI Key LYBKPDDZTNUNNM-UHFFFAOYSA-N
Synonym n-isopropylbenzylamine, n-benzylisopropylamine, isopropylbenzylamine, n-benzyl-n-isopropylamine, benzylisopropylamine, benzenemethanamine, n-1-methylethyl, n-benzyl isopropylamine, benzylamine, n-isopropyl, n-1-methylethyl-benzenemethanamine, benzyl propan-2-yl amine
PubChem CID 66024
IUPAC Name N-benzylpropan-2-amine
SMILES CC(C)NCC1=CC=CC=C1

Specifications

Density 0.91
Boiling Point 199°C to 201°C
Flash Point 87°C (190°F)
Refractive Index 1.502
Quantity 100 g
Formula Weight 149.24
Percent Purity 97%
Chemical Name or Material N-Isopropylbenzylamine
Hazard Category H227-H302-H314
Hazard Statement GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Precautionary Statement P210-P260-P264b-P270-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
DOTInformation Hazard Class: 8; Packaging Group: III
EINECSNumber 203-067-6
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.