Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Methylmorpholine, 99%, ACROS Organics™

Click to view available options
Quantity:
100g
500g
2.5kg
Packaging:
Glass Bottle
Plastic drum
109-02-4
C5H11NO
101.15
MFCD00006175
SJRJJKPEHAURKC-UHFFFAOYSA-N
n-methylmorpholine,morpholine, 4-methyl,methylmorpholine,1-methylmorpholine,morpholine, n-methyl,4-methylmorfolin,4-methylmorpholin,n-methyl morpholine,4-methylmorfolin czech,n-methylmorpholin
7972
4-methylmorpholine
CN1CCOCC1
109-02-4
C5H11NO
101.15
MFCD00006175
SJRJJKPEHAURKC-UHFFFAOYSA-N
n-methylmorpholine,morpholine, 4-methyl,methylmorpholine,1-methylmorpholine,morpholine, n-methyl,4-methylmorfolin,4-methylmorpholin,n-methyl morpholine,4-methylmorfolin czech,n-methylmorpholin
7972
4-methylmorpholine
CN1CCOCC1
Viscosity 0.91 mPa.s (20°C)
Specific Gravity 0.92
Water 0.5% max. (K.F.)
Refractive Index 1.4339 to 1.4359
Packaging Glass Bottle
Infrared Spectrum Authentic
Boiling Point 115.0°C to 116.0°C (750.0 mmHg)
Density 0.9200g/mL
Flash Point 14°C
Quantity 100g
Melting Point -65.0°C
Color Undesignated
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Beilstein 27,6
Fieser 01,690; 02,278
Merck Index 15,6362
Solubility Information Solubility in water: >500g/L (20°C). Other solubilities: miscible with alcohol,ether and benzene
Formula Weight 101.15
Physical Form Liquid
Percent Purity 99%
Chemical Name or Material N-Methylmorpholine, 99%
Show More Show Less

GHS H Statement:
Causes severe skin burns and eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Highly flammable liquid and vapor.

GHS P Statement:
Ground/Bond container and receiving equipment.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger

EINECSNumber : 203-640-0

RTECSNumber : QE5775000

TSCA : TSCA