Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Methylmorpholine (Peptide Synthesis), Fisher BioReagents

Catalog No. bp610500 Shop All Fisherbrand Products
Click to view available options
Quantity:
500 mL
Packaging:
Glass Bottle
N-Methylmorpholine is used in peptide synthesis protocols.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 109-02-4
Molecular Formula C5H11NO
Molecular Weight (g/mol) 101.15
MDL Number MFCD00006175
InChI Key SJRJJKPEHAURKC-UHFFFAOYSA-N
Synonym n-methylmorpholine, morpholine, 4-methyl, methylmorpholine, 1-methylmorpholine, morpholine, n-methyl, 4-methylmorfolin, 4-methylmorpholin, n-methyl morpholine, 4-methylmorfolin czech, n-methylmorpholin
PubChem CID 7972
IUPAC Name 4-methylmorpholine
SMILES CN1CCOCC1

Specifications

Alpha Vector Methylmorpholine
Boiling Range 2 max.
Melting Point -65°C
Color Colorless
Density 0.920g/cm³
pH 10.6
Boiling Point 115°C
Assay Percent Range ≥99.0 %
Packaging Glass Bottle
Quantity 500 mL
Merck Index 15, 6362
Vapor Pressure 22.5mmHg at 20°C
Formula Weight 101.15
Percent Purity ≥99.0%
Grade Peptide Synthesis
Physical Form Liquid
Chemical Name or Material N-Methylmorpholine
Show More Show Less
Hazard Statement DANGER!
Precautionary Statement Emergency Overview
Flammable liquid and vapor. Harmful by inhalation, in contact with skin and if swallowed. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Immediate medical attention is required. Move to fresh air. If breathing is difficult, give oxygen. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Immediate medical attention is required.

NFPA

Health:3
Flammability:3
Instability:0
Recommended Storage RT
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.