Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N myc interactor Antibody (PCRP-NMI-1C1), DyLight 405, Novus Biologicals™
SDP

Catalog No. NB390434 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB390434 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB390434 Supplier Novus Biologicals Supplier No. NBP308812V
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

N myc interactor Monoclonal antibody specifically detects N myc interactor in Human samples. It is validated for Flow Cytometry, Protein Array

Specifications

Antigen N myc interactor
Applications Flow Cytometry, Protein Array
Classification Monoclonal
Clone PCRP-NMI-1C1
Conjugate DyLight 405
Formulation 50mM Sodium Borate
Gene Alias Nmi, N-myc (and STAT) interactor, N-myc and STAT interactor, N-myc interactor, N-myc-interactor
Host Species Mouse
Immunogen Recombinant full-length human N myc interactor protein (Uniprot: Q13287)
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cancer, Signal Transduction, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 9111
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.