Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,N-Bis[2-(p-tolylsulfonyloxy)ethyl]-p-toluenesulfonamide 98.0+%, TCI America™
SDP

Catalog No. B41365G Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
5 g
25 g

Chemical Identifiers

CAS 16695-22-0
Molecular Formula C25H29NO8S3
Molecular Weight (g/mol) 567.686
MDL Number MFCD00026000
InChI Key VJDZYMIEDBLSDT-UHFFFAOYSA-N
Synonym N,N-Bis[2-(tosyloxy)ethyl]-p-toluenesulfonamide
PubChem CID 267409
IUPAC Name 2-[(4-methylphenyl)sulfonyl-[2-(4-methylphenyl)sulfonyloxyethyl]amino]ethyl 4-methylbenzenesulfonate
SMILES CC1=CC=C(C=C1)S(=O)(=O)N(CCOS(=O)(=O)C2=CC=C(C=C2)C)CCOS(=O)(=O)C3=CC=C(C=C3)C

Specifications

Melting Point 95°C
Color White
Quantity 5 g
Formula Weight 567.69
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material N,N-Bis[2-(p-tolylsulfonyloxy)ethyl]-p-toluenesulfonamide
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.