Learn More
N,N'-Bis(salicylidene)ethylenediaminecobalt(II), 96%
Description
Salcomine catalyzes the oxidation of 2,6-disubstituted phenols by dioxygen. Experimental inhibitor of human cytμloviris proteinase activity. Metal complex dyes used as redox catalysts.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Color | Black |
Odor | Odorless |
Linear Formula | [-CH2N=CH(C6H4O-)]2Co |
Quantity | 25 g |
Solubility Information | Insoluble in water. Soluble in benzene,chloroform,and pyridine. |
IUPAC Name | λ²-cobalt(2+) 2-{[(2-{[(2-oxidophenyl)methylidene]amino}ethyl)imino]methyl}benzen-1-olate |
Formula Weight | 325.23 |
Percent Purity | 96% |
Physical Form | Powder |
Chemical Name or Material | N,N'-Bis (salicylidene)ethylenediaminecobalt (II) |
Safety and Handling
Hazard Category | H302+H332-H317-H350 |
Hazard Statement | GHS H Statement H302-H332-H317 Harmful if swallowed. Harmful if inhaled. May cause an allergic skin reaction. |
Precautionary Statement | P201-P202-P261-P264b-P270-P271-P272-P280g-P281-P301+P312-P302+P352-P304+P340-P308+P313-P330-P333+P313-P363-P501c |
EINECSNumber | 238-012-5 |
RTECSNumber | GG0590000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.