Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,N'-Dibenzylethylenediamine, 97%

Catalog No. aal0354014
Click to view available options
Quantity:
25 g
100 g

CAS: 140-28-3 | C16H20N2 | 240.35 g/mol

N,N'-Dibenzylethylenediamine is an intermediate of cephalosporin and penicillin.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N,N′-Dibenzylethylenediamine is an intermediate of cephalosporin and penicillin.

Solubility
Slightly soluble in water.

Notes
Air sensitive. Store under inert gas. Store away from oxidizing agents, air.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 140-28-3
Molecular Formula C16H20N2
Molecular Weight (g/mol) 240.35
MDL Number MFCD00004771
InChI Key JUHORIMYRDESRB-UHFFFAOYSA-N
Synonym n,n'-dibenzylethylenediamine, benzathine, dbed, benzatin, n1,n2-dibenzylethane-1,2-diamine, 1,2-bis benzylamino ethane, 1,2-ethanediamine, n,n'-bis phenylmethyl, usaf do-53, unii-c659vz7p7t, ethylenediamine, n,n'-dibenzyl
PubChem CID 8793
ChEBI CHEBI:51344
IUPAC Name N,N'-dibenzylethane-1,2-diamine
SMILES C1=CC=C(C=C1)CNCCNCC2=CC=CC=C2

Specifications

Melting Point 23°C to 25°C
Density 1.025
Boiling Point 184°C to 186°C (2 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.565
Linear Formula C6H5CH2NHCH2CH2NHCH2C6H5
Quantity 25 g
UN Number UN2735
Beilstein 786668
Sensitivity Air Sensitive
Merck Index 14,1064
Solubility Information Slightly soluble in water.
Formula Weight 240.35
Percent Purity 97%
Chemical Name or Material N,N'-Dibenzylethylenediamine
Show More Show Less
Hazard Category H302-H314-H335
Hazard Statement GHS H Statement
H301-H314-H318
Toxic if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
EINECSNumber 205-408-4
RTECSNumber KV3325000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.