Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,N-Diethylmethylamine, 98%, Thermo Scientific Chemicals

Catalog No. AC114465000
Click to view available options
Quantity:
500 mL
Packaging:
Glass Bottle

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 616-39-7
Molecular Formula C5H14N
Molecular Weight (g/mol) 88.17
MDL Number MFCD00009049
InChI Key GNVRJGIVDSQCOP-UHFFFAOYSA-O
Synonym n,n-diethylmethylamine, diethylmethylamine, ethanamine, n-ethyl-n-methyl, n-methyldiethylamine, diethylamine, n-methyl, n-ethyl-n-methyl-ethanamine, methyldiethylamine, diethylmethyl amine, diethyl methyl amine, methyl diethyl amine
PubChem CID 12022
IUPAC Name N-ethyl-N-methylethanamine
SMILES CC[NH+](C)CC

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Colorless to Yellow
Density 0.7200g/mL
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass Bottle
Refractive Index 1.3890 to 1.3910 (20°C, 589nm)
Linear Formula (C2H5)2NCH3
Quantity 500 mL
Beilstein 04, 99
Formula Weight 87.15
Percent Purity ≥97.5%
Physical Form Liquid
Chemical Name or Material N, N-Diethylmethylamine, 98%
Show More Show Less
Hazard Category Danger
Hazard Statement GHS H Statement:
Causes serious eye damage.
Harmful if inhaled.
Harmful if swallowed.
Causes severe skin burns and eye damage.
Highly flammable liquid and vapor.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 210-480-5
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.