Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
N,N-Dimethylformamide-d{7}, 99.5% (Isotopic)
Click to view available options
Quantity:
1 g
5 g
10 g
5 x 1 g
Description
It is a common solvent used in NMR spectroscopy. It is also used as a reagent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Physical Form | Liquid |
| Color | Colorless |
| Quantity | 5 g |
| Packaging | Prescored Ampule |
| Flash Point | 58°C (136°F) |
| Density | 1.04 g/mL |
| Odor | Amine-like |
| Boiling Point | 153°C |
| Linear Formula | DCON(CD3)2 |
| UN Number | UN2265 |
| Show More |
Safety and Handling
| Hazard Category | H226-H312+H332-H319-H360D |
| Hazard Statement | GHS H Statement H360-H226-H312-H332-H319 May damage fertility or the unborn child. Flammable liquid and vapor. Harmful in contact with skin. Harmful if inhaled. Causes serious eye irritation. |
| Precautionary Statement | P201-P202-P210-P233-P240-P241-P242-P243-P261-P264b-P271-P280i-P281-P303+P361+P353-P304+P340-P305+P351+P338-P308+P313-P312-P363-P370+P378q-P501c |
| DOTInformation | Transport Hazard Class: 3; Packaging Group: III; Proper Shipping Name: N,N-DIMETHYLFORMAMIDE |
| EINECSNumber | 224-745-8 |
| TSCA | No |
| Recommended Storage | Ambient |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction