Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,N'-Disuccinimidyl carbonate, tech. 85%, remainder N-Hydroxysuccinimide

Catalog No. AAH2662906
Click to view available options
Quantity:
1 g
5 g
25 g

CAS: 74124-79-1 | C9H8N2O7 | 256.17 g/mol

N,N'-Disuccinimidyl carbonate is used to prepare cleavable linker succinimide vinylsufonylethyl carbonate for thiol modified-DNA. It serves as a reagent for the synthesis of carbamates. Further, it is used as a coupling reagent as well as in the preparation of N-succinimidyl esters of N-protected amino acids, activated carbonate and coupling of ligand to proteins.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N,N′-Disuccinimidyl carbonate is used to prepare cleavable linker succinimide vinylsufonylethyl carbonate for thiol modified-DNA. It serves as a reagent for the synthesis of carbamates. Further, it is used as a coupling reagent as well as in the preparation of N-succinimidyl esters of N-protected amino acids, activated carbonate and coupling of ligand to proteins.

Solubility
Soluble in dimethyl sulfoxide, hot pyridine, acetonitrile and most organic solvents. Insoluble in water.

Notes
Moisture sensitive. Store in a cool place. Incompatible with strong acids and oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 74124-79-1
Molecular Formula C9H8N2O7
Molecular Weight (g/mol) 256.17
MDL Number MFCD00009767
InChI Key PFYXSUNOLOJMDX-UHFFFAOYSA-N
Synonym n,n'-disuccinimidyl carbonate, bis 2,5-dioxopyrrolidin-1-yl carbonate, n,n'-disuccinimidylcarbonate, di succinimido carbonate, disuccinimidyl carbonate, di n-succinimidyl carbonate, n,n-disuccinimidyl carbonate, dsc, disuccinimido carbonate, pubchem12700
PubChem CID 676246
IUPAC Name bis(2,5-dioxopyrrolidin-1-yl) carbonate
SMILES C1CC(=O)N(C1=O)OC(=O)ON2C(=O)CCC2=O

Specifications

Melting Point 190°C (decomposition)
Quantity 5 g
Beilstein 1499137
Sensitivity Hygroscopic
Solubility Information Soluble in dimethyl sulfoxide,hot pyridine,acetonitrile and most organic solvents. Insoluble in water.
Formula Weight 256.17
Percent Purity 85%
Grade Technical
Assay remainder N-Hydroxysuccinimide
Chemical Name or Material N,N'-Disuccinimidyl carbonate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 277-730-3
TSCA No
Recommended Storage Keep cold; Store under Nitrogen

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.