Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,N'-Hexamethylenebisacetamide, 98%, Thermo Scientific™

Catalog No. AC208322500
Click to view available options
Quantity:
5 g
50 g
250 g
Packaging:
Glass bottle

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3073-59-4
Molecular Formula C10H20N2O2
Molecular Weight (g/mol) 200.28
MDL Number MFCD00008684
InChI Key BNQSTAOJRULKNX-UHFFFAOYSA-N
Synonym hmba, hexamethylene bisacetamide, hexamethylenebisacetamide, n,n'-hexamethylenebisacetamide, hexamethylenediacetamide, n,n'-diacetyl-1,6-diaminohexane, diacetyldiaminohexane, n,n'-diacetylhexamethylenediamine, n,n'-hexane-1,6-diyl diacetamide, acetamide, n,n'-hexamethylenebis
PubChem CID 3616
IUPAC Name N-(6-acetamidohexyl)acetamide
SMILES CC(=O)NCCCCCCNC(C)=O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 126°C to 129°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula CH3CONH(CH2)6NHCOCH3
Quantity 250 g
Beilstein 04, 269
Formula Weight 200.28
Percent Purity ≥97.5%
Physical Form Crystalline Powder or Flakes
Chemical Name or Material N, N'-Hexamethylenebisacetamide
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.