Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
N,N,N',N″,N″-Pentamethyldiethylenetriamine 99.0+%, TCI America™
Shop All Tokyo Chemical Industry (TCI) Products

Click to view available options
Quantity:
25 mL
100 mL
500 mL
Specifications
Specifications
| Melting Point | -20°C |
| Color | Yellow |
| Boiling Point | 86°C |
| Quantity | 100 mL |
| UN Number | 2922 |
| Formula Weight | 173.30 |
| Percent Purity | ≥99.0% (GC) |
| Physical Form | Liquid |
| Chemical Name or Material | N,N,N′,N′′,N′′-Pentamethyldiethylenetriamine |
Safety and Handling
| EINECSNumber | (2)-0147 |
| RTECSNumber | IE2100000 |
| TSCA | Yes |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction