Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,O-Bis(trimethylsilyl)trifluoroacetamide, 98+%

Catalog No. AAA1203309
Click to view available options
Quantity:
10 g
50 g
100 g
250 g

Catalog No. AAA1203309

Supplier: Thermo Scientific Chemicals A1203309

Only null left
Add to Cart

CAS: 25561-30-2 | C8H18F3NOSi2 | 257.403 g/mol

N,O-Bis(trimethylsilyl)trifluoroacetamide is used as a derivatization reagent for gas chromatography and mass-spectrometry analysis of phenolic acids in fruits. It is also used as a reactant in the preparation of pyrimidinone ribosides and analogs which shows anti-tumor properties. It plays as an important role as a reagent for the regioselective desulfation of polysaccharide sulfates. Furthermore, it is used to prepare trimethylsilyl ethers of carbohydrates and alcohols.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N,O-Bis(trimethylsilyl)trifluoroacetamide is used as a derivatization reagent for gas chromatography and mass-spectrometry analysis of phenolic acids in fruits. It is also used as a reactant in the preparation of pyrimidinone ribosides and analogs which shows anti-tumor properties. It plays as an important role as a reagent for the regioselective desulfation of polysaccharide sulfates. Furthermore, it is used to prepare trimethylsilyl ethers of carbohydrates and alcohols.

Solubility
Miscible with chloroform.

Notes
Moisture sensitive. Store in cool place. Keep the container tightly closed in a dry and well-ventilated place Incompatible with strong oxidizing agents and strong acids.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25561-30-2
Molecular Formula C8H18F3NOSi2
MDL Number MFCD00008269
Synonym bstfa, n,o-bis trimethylsilyl trifluoroacetamide, acetamide, 2,2,2-trifluoro-o,n-bis trimethylsilyl, trimethylsilyl 2,2,2-trifluoro-n-trimethylsilyl acetimidate, trimethylsilyl 2,2,2-trifluoro-n-trimethylsilyl ethanimidate, trimethylsilyl 1z-2,2,2-trifluoro-n-z-trimethylsilyl ethanimidoate #, n,o-bis trimethylsilyl trifluoroacetamide bstfa, trimethylsilyl 2,2,2-trifluoro-n-trimethylsilyl ethanecarboximidate
PubChem CID 9601896

Specifications

Melting Point -10°C
Density 0.97
Boiling Point 145°C to 147°C
Flash Point 23°C (73°F)
Refractive Index 1.384
Quantity 10 g
UN Number UN2924
Beilstein 3607620
Sensitivity Moisture sensitive
Solubility Information Miscible with chloroform.
InChI Key XCOBLONWWXQEBS-GHXNOFRVSA-N
SMILES C[Si](C)(C)N=C(C(F)(F)F)O[Si](C)(C)C
IUPAC Name trimethylsilyl (1Z)-2,2,2-trifluoro-N-trimethylsilylethanimidate
Molecular Weight (g/mol) 257.403
Formula Weight 257.4
Percent Purity ≥98%
Chemical Name or Material N,O-Bis (trimethylsilyl)trifluoroacetamide
Show More Show Less
Hazard Category H226-H314-H335
Hazard Statement GHS H Statement
H314-H318-H226
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapor.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, CORROSIVE, N.O.S.
EINECSNumber 247-103-9
TSCA No
Recommended Storage Keep cold; Store under Nitrogen

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.