Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

n-Octacosane, 99%

Catalog No. p-7052060
Click to view available options
Quantity:
10 g
50 g
250 g

CAS: 630-02-4 | C28H58 | 394.77 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 630-02-4
Molecular Formula C28H58
Molecular Weight (g/mol) 394.77
MDL Number MFCD00009355
InChI Key ZYURHZPYMFLWSH-UHFFFAOYSA-N
Synonym n-octacosane, n-octcosane, unii-vff49836p8, ch3-ch2 26-ch3, octacosane, analytical standard, ccris 680, octacosane, n, octacosane, heptacosane, methyl, n-octacosane 10g
PubChem CID 12408
ChEBI CHEBI:32943
IUPAC Name octacosane
SMILES CCCCCCCCCCCCCCCCCCCCCCCCCCCC

Specifications

Melting Point 61°C to 63°C
Boiling Point 440°C
Flash Point 227°C (441°F)
Quantity 10 g
Solubility Information Soluble in chloroform,acetone,benzene,and toluene. Insoluble in water.
Formula Weight 394.77
Percent Purity 99%
Physical Form Crystalline
Chemical Name or Material n-Octacosane
EINECSNumber 211-125-7
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.