Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

n-Octadecane, 99%

Catalog No. aa3195422
Click to view available options
Quantity:
100 g
500 g
2 kg

CAS: 593-45-3 | C18H38 | 254.502 g/mol

n-octadecane is used as solvent, lubricant, transformer oil and anti-corrosion agents. It is also used in paraffin as well as chemical intermediate in organic synthesis. It plays an important role in pheromones as a chemical messenger. N-Octadecane/ molecular sieve 5A composites find application as thermal storage material.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 593-45-3
Molecular Formula C18H38
Molecular Weight (g/mol) 254.502
MDL Number MFCD00009007
InChI Key RZJRJXONCZWCBN-UHFFFAOYSA-N
Synonym n-octadecane, octadecan, unii-n102p6haiu, n102p6haiu, octadecane, octadecane, analytical standard, octadecane, n, oktadekan, ccris 681, stearyl group
PubChem CID 11635
ChEBI CHEBI:32926
IUPAC Name octadecane
SMILES CCCCCCCCCCCCCCCCCC

Specifications

Melting Point 28°C
Density 0.782 g/mL
Boiling Point 317°C
Flash Point 166°C (330°F)
Assay Percent Range 99%
Refractive Index 1.439
Quantity 100 g
Solubility Information Miscible with alcohol,acetone,ether and petroleum ether. Immiscible with water.
Formula Weight 254.5
Percent Purity 99%
Physical Form Crystalline Mass/Melt
Chemical Name or Material n-Octadecane
Show More Show Less
Hazard Category H304-H315-H319-H335
Hazard Statement GHS H Statement
H304
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 209-790-3
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.