Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

n-Octane, 99+%, extra pure

Catalog No. p-3737102
Click to view available options
Quantity:
2 mL
500 mL
2.5 L
25 L
Packaging:
Glass Bottle
Metal drum
This item is not returnable. View return policy
This item is not returnable. View return policy

CAS: 111-65-9 | C8H18 | 114.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 111-65-9
Molecular Formula C8H18
Molecular Weight (g/mol) 114.23
MDL Number MFCD00009556
InChI Key TVMXDCGIABBOFY-UHFFFAOYSA-N
Synonym n-octane, oktan, oktanen, ottani, oktanen dutch, heptane, methyl, oktan polish, ottani italian, octane, all isomers, unii-x1rv0b2fjv
PubChem CID 356
ChEBI CHEBI:17590
IUPAC Name octane
SMILES CCCCCCCC

Specifications

CAS Min % 99.0
CAS Max % 100.0
Melting Point -57.0°C
Color Colorless
Density 0.7080g/mL
Boiling Point 125.0°C to 127.0°C
Flash Point 13°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Glass Bottle
Refractive Index 1.3965 to 1.3985
Linear Formula CH3(CH2)6CH3
Quantity 2 mL
Residue after Evaporation 0.05% max.
Beilstein 01, 159
Specific Gravity 0.708
Merck Index 15, 6838
Solubility Information Solubility in water: 0.0007g/L (20°C). Other solubilities: soluble in alcohol and acetone, freely miscible with ether, benzene, petroleum, ether and gasoline
Viscosity 0.55 mPa.s (20°C)
Vapor Pressure 14mbar at 20°C
Formula Weight 114.23
Percent Purity 99+%
Grade Extra Pure
Physical Form Liquid
Chemical Name or Material n-Octane
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes skin irritation.
Very toxic to aquatic life with long lasting effects.
May cause drowsiness or dizziness.
May be fatal if swallowed and enters airways.
Highly flammable liquid and vapor.
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician.
Do NOT induce vomiting.
Avoid release to the environment.
IF ON SKIN: Wash with plenty of soap
EINECSNumber 203-892-1

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.