Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-PAC Antibody, Alexa Fluor™ 647, Novus Biologicals™
SDP

Catalog No. NB084906 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB084906 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB084906 Supplier Novus Biologicals Supplier No. NBP299354AF647
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

N-PAC Polyclonal specifically detects N-PAC in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen N-PAC
Applications Western Blot, Immunofluorescence
Classification Polyclonal
Conjugate Alexa Fluor 647
Formulation 50 mM Sodium Borate
Gene Alias BM045, Cytokine-like nuclear factor N-PAC, Glyoxylate reductase 1 homolog, glyoxylate reductase 1 homolog (Arabidopsis), HIBDLnuclear protein 60 kDa, NP603-hydroxyisobutyrate dehydrogenase-like protein, N-PAC, nuclear protein 60kDa, Nuclear protein NP60, Nuclear protein of 60 kDa, putative oxidoreductase GLYR1
Host Species Rabbit
Immunogen Produced in rabbits immunized with a synthetic peptide corresponding to the center region of human N-PAC.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84656
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.