Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

n-Pentacosane, MP Biomedicals

Catalog No. ICN225426
Click to view available options
Quantity:
1 g

Chemical Identifiers

CAS 629-99-2
Molecular Formula C25H52
Molecular Weight (g/mol) 352.691
InChI Key YKNWIILGEFFOPE-UHFFFAOYSA-N
Synonym n-pentacosane, unii-bon9h94y8v, bon9h94y8v, pentacosane, analytical standard, pentacosane, tetracosane, methyl, n-pentacosane 1g, pentacosane standardmaterialforgc, ch3-ch2 23-ch3, solution, 10,000 mg/l, 1 ml rm, iso guide 34
PubChem CID 12406
ChEBI CHEBI:32938
IUPAC Name pentacosane
SMILES CCCCCCCCCCCCCCCCCCCCCCCCC

Specifications

Quantity 1 g
Formula Weight 352.69
Chemical Name or Material n-Pentacosane
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.