Learn More
n-Pentadecane, 99%
Description
n-pentadecane is used as a reference substance for gas chromatography. It is also used as standard materials for the determination of combustion quality of diesel. It is involved in the organic synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
n-pentadecane is used as a reference substance for gas chromatography. It is also used as standard materials for the determination of combustion quality of diesel. It is involved in the organic synthesis.
Solubility
Immiscible with water.
Notes
Incompatible with strong oxidizing agents.

Specifications
Specifications
Melting Point | 9°C to 10°C |
Density | 0.769 |
Boiling Point | 269°C to 270°C |
Flash Point | 132°C (269°F) |
Refractive Index | 1.432 |
Quantity | 250 g |
Beilstein | 1698194 |
Solubility Information | Immiscible with water. |
Formula Weight | 212.42 |
Percent Purity | 99% |
Show More |
Safety and Handling
Hazard Category | H304 |
Hazard Statement | GHS H Statement H304 May be fatal if swallowed and enters airways. |
Precautionary Statement | P301+P310-P331-P501c |
EINECSNumber | 211-098-1 |
RTECSNumber | RZ1800000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.