Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

n-Pentadecane 99%, ACROS Organics™

Click to view available options
Quantity:
25mL
250mL
1L
Packaging:
Glass bottle
629-62-9
C15H32
212.42
MFCD00008990
YCOZIPAWZNQLMR-UHFFFAOYSA-N
n-pentadecane,pentadekan,unii-16h6k2s8m2,ch3-ch2 13-ch3,mys,pentadecane, analytical standard,pentadecan,medicinal plant,pentadecyl group,pentadecane, n
12391
CHEBI:28897
pentadecane
CCCCCCCCCCCCCCC
629-62-9
C15H32
212.42
MFCD00008990
YCOZIPAWZNQLMR-UHFFFAOYSA-N
n-pentadecane,pentadekan,unii-16h6k2s8m2,ch3-ch2 13-ch3,mys,pentadecane, analytical standard,pentadecan,medicinal plant,pentadecyl group,pentadecane, n
12391
CHEBI:28897
pentadecane
CCCCCCCCCCCCCCC
Flash Point 132°C
Color Undesignated
Infrared Spectrum Authentic
Quantity 25mL
Melting Point 9.9°C
Density 0.7730g/mL
Packaging Glass bottle
Boiling Point 270.0°C
Specific Gravity 0.773
Refractive Index 1.4307 to 1.4327
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Linear Formula CH3(CH2)13CH3
Beilstein 01, 172
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohol
Formula Weight 212.42
Physical Form Liquid
Percent Purity 99%
Chemical Name or Material n-Pentadecane, 99%
Show More Show Less

GHS H Statement:
May be fatal if swallowed and enters airways.
Repeated exposure may cause skin dryness or cracking.

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician.
Do NOT induce vomiting.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 211-098-1

RTECSNumber : RZ1800000

TSCA : TSCA