Learn More
n-Pentane, 98%
Description
n-Pentane is used in consumer merchandise such as spot lifters/cleaners and foaming shave gels and in commercial products such as a blowing agent for expanded polystyrene (foam) insulation. It may also be used as a solvent (in pure form) in laboratory, chemical analysis, and other settings where a highly volatile non-polar solvent is needed.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | -130°C |
| Color | Colorless |
| Density | 0.626 g/mL |
| Boiling Point | 36°C |
| Quantity | 250 mL |
| Flash Point | -49°C (-57°F) |
| Odor | Nearly odorless |
| Physical Form | Liquid |
| Refractive Index | 1.358 |
| Assay Percent Range | 98% |
| Show More |
Safety and Handling
| Hazard Category | H225-H304-H336 |
| Hazard Statement | GHS H Statement H225-H304-H336 Highly flammable liquid and vapor. May be fatal if swallowed and enters airways. May cause drowsiness or dizziness. |
| Precautionary Statement | P210-P233-P235-P240-P241-P242-P243-P261-P271-P280-P301+P310-P303+P361+P353-P304+P340-P312-P331-P370+P378q-P501c |
| DOTInformation | Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: PENTANES |
| EINECSNumber | 203-692-4 |
| RTECSNumber | RZ9450000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.