Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

n-Pentane, 98%

Catalog No. p-7049649
Click to view available options
Quantity:
250 mL
1 L
4 L

Catalog No. AA17381AK

Supplier: Thermo Scientific Chemicals 017381AK

Only null left
Add to Cart

CAS: 109-66-0 | C5H12 | 72.15 g/mol

n-Pentane is used in consumer merchandise such as spot lifters/cleaners and foaming shave gels and in commercial products such as a blowing agent for expanded polystyrene (foam) insulation. It may also be used as a solvent (in pure form) in laboratory, chemical analysis, and other settings where a highly volatile non-polar solvent is needed.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 109-66-0
Molecular Formula C5H12
Molecular Weight (g/mol) 72.15
MDL Number MFCD00009498
InChI Key OFBQJSOFQDEBGM-UHFFFAOYSA-N
Synonym n-pentane, pentan, skellysolve a, pentanen, pentani, amyl hydride, tetrafume, tetrakil, tetraspot, pentan polish
PubChem CID 8003
ChEBI CHEBI:37830
IUPAC Name pentane
SMILES CCCCC

Specifications

Melting Point -130°C
Color Colorless
Density 0.626 g/mL
Boiling Point 36°C
Quantity 250 mL
Flash Point -49°C (-57°F)
Odor Nearly odorless
Physical Form Liquid
Refractive Index 1.358
Assay Percent Range 98%
Linear Formula CH3(CH2)3CH3
UN Number UN1265
Merck Index 14,7116
Solubility Information Miscible with water.
Formula Weight 72.15
Percent Purity 98%
Chemical Name or Material n-Pentane
Show More Show Less
Hazard Category H225-H304-H336
Hazard Statement GHS H Statement
H225-H304-H336
Highly flammable liquid and vapor.
May be fatal if swallowed and enters airways.
May cause drowsiness or dizziness.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P271-P280-P301+P310-P303+P361+P353-P304+P340-P312-P331-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: PENTANES
EINECSNumber 203-692-4
RTECSNumber RZ9450000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.